accessibility menu, dialog, popup

UserName

CH3OH3CO(Z)

CAS RN 2497-18-9

IUPAC Name: (2Z)-hex-2-en-1-yl acetate
Synonyms: trans-2-hexenyl acetate
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: HRHOWZHRCRZVCU-WAYWQWQTSA-N
SMILES: CCC\C=C/COC(C)=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)