accessibility menu, dialog, popup

UserName

CAS RN 2495-39-8

NaOSOOH2C

CAS RN 2495-39-8

IUPAC Name: sodium prop-2-ene-1-sulfonate
Synonyms: sodium prop-2-ene-1-sulfonate
Molecular Weight (g/mol): 144.12
Molecular Formula: C3H5NaO3S
InChi Key: DIKJULDDNQFCJG-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results