CAS RN 2491-37-4
IUPAC Name:
2-bromo-1-(3-hydroxyphenyl)ethan-1-one
Synonyms:
2-bromo-1-(3-hydroxyphenyl)ethanone
Molecular Weight (g/mol):
215.05
Molecular Formula:
C8H7BrO2
InChi Key:
IEPSGFQQGKPTPM-UHFFFAOYSA-N
SMILES:
OC1=CC=CC(=C1)C(=O)CBr
1
–
1
of
1
results
1
Promotions Available
2-Bromo-3'-hydroxyacetophenone, 96%
2-Bromo-3'-hydroxyacetophenone, 96%
CAS: 2491-37-4 Molecular Formula: C8H7BrO2 Molecular Weight (g/mol): 215.04 MDL Number: MFCD05664326 InChI Key: IEPSGFQQGKPTPM-UHFFFAOYSA-N PubChem CID: 6917206 IUPAC Name: 2-bromo-1-(3-hydroxyphenyl)ethanone SMILES: C1=CC(=CC(=C1)O)C(=O)CBr