accessibility menu, dialog, popup

UserName

CAS RN 24892-49-7

H3CCH3HOCH3CH3HO

CAS RN 24892-49-7

IUPAC Name: 2,4-dimethylpentane-2,4-diol
Synonyms: 2,4-dimethylpentane-2,4-diol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: DBTGFWMBFZBBEF-UHFFFAOYSA-N
SMILES: CC(C)(O)CC(C)(C)O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results