accessibility menu, dialog, popup

UserName

H3COClCl

CAS RN 2476-37-1

IUPAC Name: 1-(2,5-dichlorophenyl)ethan-1-one
Synonyms: 1-(2,5-dichlorophenyl)ethanone
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: CYNFEPKQDJHIMV-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=CC=C1Cl
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)