accessibility menu, dialog, popup

UserName

CAS RN 2474-07-9

IUPAC Name: [2-(4-bromophenoxy)ethyl]dimethylamine
Synonyms: [2-(4-bromophenoxy)ethyl]dimethylamine
Molecular Weight (g/mol): 244.13
Molecular Formula: C10H14BrNO
InChi Key: MOVOYJFCKMYLHQ-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results