accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 24731-17-7

IUPAC Name: ethyl 2-(2-oxocyclohexyl)acetate
Synonyms: ethyl 2-(2-oxocyclohexyl)acetate
Molecular Weight (g/mol): 184.24
Molecular Formula: C10H16O3
InChi Key: ZZWSNYNCRUZSPR-UHFFFAOYNA-N
SMILES: CCOC(=O)CC1CCCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)