accessibility menu, dialog, popup

UserName

CH3OBOH3COCH3

CAS RN 2467-12-1

IUPAC Name: trioctyl borate
Synonyms: boric acid (H3BO3), trioctyl ester
Molecular Weight (g/mol): 398.48
Molecular Formula: C24H51BO3
InChi Key: DTBRTYHFHGNZFX-UHFFFAOYSA-N
SMILES: CCCCCCCCOB(OCCCCCCCC)OCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results