accessibility menu, dialog, popup

UserName

BFFFFCH3NNH3C

CAS RN 244193-48-4

IUPAC Name: 3-methyl-1-propyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1-methyl-3-propylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol): 212
Molecular Formula: C7H13BF4N2
InChi Key: XFTQQJUVYZPNRC-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results