accessibility menu, dialog, popup

UserName

CAS RN 24404-53-3

H3COOOSOOCH3OOCH3OOH3C

CAS RN 24404-53-3

IUPAC Name: [3,4,5-tris(acetyloxy)-6-(phenylsulfanyl)oxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-(phenylsulfanyl)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 440.46
Molecular Formula: C20H24O9S
InChi Key: JCKOUAWEMPKIAT-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(SC2=CC=CC=C2)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results