accessibility menu, dialog, popup

UserName

H3CN

CAS RN 2437-25-4

IUPAC Name: dodecanenitrile
Synonyms: lauronitrile
Molecular Weight (g/mol): 181.32
Molecular Formula: C12H23N
InChi Key: VXCUURYYWGCLIH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results