CAS RN 243643-97-2
IUPAC Name:
2-(2-chloroethanesulfonyl)-5-(trifluoromethyl)pyridine
Synonyms:
2-(2-chloroethanesulfonyl)-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol):
273.65
Molecular Formula:
C8H7ClF3NO2S
InChi Key:
VACOIIVHOIMMEF-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CN=C(C=C1)S(=O)(=O)CCCl
1
–
1
of
1
results
1
2-[(2-chloroethyl)sulfonyl]-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
CAS: 243643-97-2 Molecular Formula: C8H7ClF3NO2S Molecular Weight (g/mol): 273.654 MDL Number: MFCD00099609 InChI Key: VACOIIVHOIMMEF-UHFFFAOYSA-N Synonym: 2-2-chloroethyl sulfonyl-5-trifluoromethyl pyridine,2-2-chloroethanesulfonyl-5-trifluoromethyl pyridine,2-2-chloroethylsulfonyl-5-trifluoromethyl pyridine,2-2-chloroethyl sulphonyl-5-trifluoromethyl pyridine,pyridine,2-2-chloroethyl sulfonyl-5-trifluoromethyl PubChem CID: 2779640 IUPAC Name: 2-(2-chloroethylsulfonyl)-5-(trifluoromethyl)pyridine SMILES: C1=CC(=NC=C1C(F)(F)F)S(=O)(=O)CCCl