accessibility menu, dialog, popup

UserName

CAS RN 24302-67-8

IUPAC Name: hydrogen (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol chloride
Synonyms: hydrogen (+)-cinconine chloride
Molecular Weight (g/mol): 330.86
Molecular Formula: C19H23ClN2O
InChi Key: IMUHWLVEEVGMBC-BKUXTCEESA-N
SMILES: [H+].[Cl-].O[C@H]([C@H]1C[C@@H]2CCN1C[C@@H]2C=C)C1=C2C=CC=CC2=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results