CAS RN 2420-87-3
IUPAC Name:
5-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms:
BPDA
Molecular Weight (g/mol):
294.22
Molecular Formula:
C16H6O6
InChi Key:
WKDNYTOXBCRNPV-UHFFFAOYSA-N
SMILES:
O=C1OC(=O)C2=CC(=CC=C12)C1=CC=C2C(=O)OC(=O)C2=C1
1
–
2
of
2
results
1
4,4'-Biphthalic Anhydride 98.0+%, TCI America™
CAS: 2420-87-3 Molecular Formula: C16H6O6 Molecular Weight (g/mol): 294.218 MDL Number: MFCD00039140 InChI Key: WKDNYTOXBCRNPV-UHFFFAOYSA-N PubChem CID: 75494 IUPAC Name: 5-(1,3-dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione SMILES: C1=CC2=C(C=C1C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
2
4,4'-Biphthalic Anhydride (purified by sublimation) 98.0+%, TCI America™
CAS: 2420-87-3 Molecular Formula: C16H6O6 Molecular Weight (g/mol): 294.218 MDL Number: MFCD00039140 InChI Key: WKDNYTOXBCRNPV-UHFFFAOYSA-N Synonym: Diphenyl-3,3′C,4,4′C-tetracarboxylic Dianhydride PubChem CID: 75494 IUPAC Name: 5-(1,3-dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione SMILES: C1=CC2=C(C=C1C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O