accessibility menu, dialog, popup

UserName

CAS RN 2419-94-5

CH3CH3CH3OOHN(S)OOOO

CAS RN 2419-94-5

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]pentanedioate
Molecular Weight (g/mol): 245.23
Molecular Formula: C10H15NO6
InChi Key: AQTUACKQXJNHFQ-LURJTMIESA-L
SMILES: CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results