accessibility menu, dialog, popup

UserName

CAS RN 2414-98-4

IUPAC Name: magnesium(2+) diethanolate
Synonyms: magnesium(2+) bis(ethoxide)
Molecular Weight (g/mol): 114.43
Molecular Formula: C4H10MgO2
InChi Key: XDKQUSKHRIUJEO-UHFFFAOYSA-N
SMILES: [Mg++].CC[O-].CC[O-]
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)