CAS RN 24019-66-7
IUPAC Name:
2-hydroxy-3-(prop-2-en-1-yl)benzaldehyde
Synonyms:
2-hydroxy-3-(prop-2-en-1-yl)benzaldehyde
Molecular Weight (g/mol):
162.19
Molecular Formula:
C10H10O2
InChi Key:
INLWEXRRMUMHKB-UHFFFAOYSA-N
SMILES:
OC1=C(CC=C)C=CC=C1C=O
1
–
1
of
1
results
1
Promotions Available
3-Allylsalicylaldehyde, 97%
3-Allylsalicylaldehyde, 97%
CAS: 24019-66-7 Molecular Formula: C10H10O2 Molecular Weight (g/mol): 162.188 MDL Number: MFCD00037364 InChI Key: INLWEXRRMUMHKB-UHFFFAOYSA-N Synonym: 3-allyl-2-hydroxybenzaldehyde,3-allylsalicylaldehyde,3-allyl-2-hydroxy-benzaldehyde,benzaldehyde, 2-hydroxy-3-2-propenyl,salicylaldehyde, 3-allyl,2-hydroxy-3-prop-2-en-1-yl benzaldehyde,acmc-209wev,tpc-sk001,2-hydroxy-3-allylbenzaldehyde PubChem CID: 141062 IUPAC Name: 2-hydroxy-3-prop-2-enylbenzaldehyde SMILES: C=CCC1=CC=CC(=C1O)C=O