accessibility menu, dialog, popup

UserName

CAS RN 23943-96-6

IUPAC Name: (2R)-2-methoxypropanoate
Synonyms: (2R)-2-methoxypropanoate
Molecular Weight (g/mol): 103.1
Molecular Formula: C4H7O3
InChi Key: ICPWFHKNYYRBSZ-GSVOUGTGSA-M
SMILES: CO[C@H](C)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results