accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 2384-70-5

IUPAC Name: dec-2-yne
Synonyms: dec-2-yne
Molecular Weight (g/mol): 138.25
Molecular Formula: C10H18
InChi Key: RWDDSTHSVISBEA-UHFFFAOYSA-N
SMILES: CCCCCCCC#CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results