accessibility menu, dialog, popup

UserName

ClH3CNCH3NNH2O

CAS RN 2381-85-3

IUPAC Name: 12-amino-8-(diethylamino)-10λ⁴-oxa-5-azatetraphen-10-ylium chloride
Synonyms: nile blue cresyl fast violet cresol fast violet nile blue dye nile blue chloride
Molecular Weight (g/mol): 353.85
Molecular Formula: C20H20ClN3O
InChi Key: XJCPMUIIBDVFDM-UHFFFAOYSA-M
SMILES: [Cl-].CCN(CC)C1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results