accessibility menu, dialog, popup

UserName

CAS RN 238088-16-9

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)butanenitrile
Molecular Weight (g/mol): 195.07
Molecular Formula: C10H18BNO2
InChi Key: LVMSRWXPZLRTIC-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCCC#N)OC1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results