accessibility menu, dialog, popup

UserName

CAS RN 23739-88-0

IUPAC Name: [36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecakis(acetyloxy)-10,15,20,25,30,35-hexakis[(acetyloxy)methyl]-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontan-5-yl]methyl acetate
Synonyms: [36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecakis(acetyloxy)-10,15,20,25,30,35-hexakis[(acetyloxy)methyl]-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontan-5-yl]methyl acetate
Molecular Weight (g/mol): 2017.76
Molecular Formula: C84H112O56
InChi Key: NOPKOJDDVCBPTP-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC2OC3C(COC(C)=O)OC(OC4C(COC(C)=O)OC(OC5C(COC(C)=O)OC(OC6C(COC(C)=O)OC(OC7C(COC(C)=O)OC(OC8C(COC(C)=O)OC(OC1C(OC(C)=O)C2OC(C)=O)C(OC(C)=O)C8OC(C)=O)C(OC(C)=O)C7OC(C)=O)C(OC(C)=O)C6OC(C)=O)C(OC(C)=O)C5OC(C)=O)C(OC(C)=O)C4OC(C)=O)C(OC(C)=O)C3OC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results