accessibility menu, dialog, popup

UserName

ONH2OHOH

CAS RN 2370-61-8

IUPAC Name: 2-amino-3-(2-hydroxyphenyl)propanoic acid
Synonyms: 2-tyrosine|o-tyrosine|ortho-tyrosine|DL-o-tyrosine|DL- O-tyrosine|o-tyrosine, DL-|DL-ortho-tyrosine
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: WRFPVMFCRNYQNR-UHFFFAOYNA-N
SMILES: NC(CC1=CC=CC=C1O)C(O)=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results