CAS RN 23384-72-7
IUPAC Name:
1-(3,4-difluorophenyl)propan-1-one
Synonyms:
1-(3,4-difluorophenyl)propan-1-one
Molecular Weight (g/mol):
170.16
Molecular Formula:
C9H8F2O
InChi Key:
FSZOBSMJJFHVCQ-UHFFFAOYSA-N
SMILES:
CCC(=O)C1=CC=C(F)C(F)=C1
1
–
1
of
1
results
1
Promotions Available
3',4'-Difluoropropiophenone, 97%
3',4'-Difluoropropiophenone, 97%
CAS: 23384-72-7 Molecular Formula: C9H8F2O Molecular Weight (g/mol): 170.16 MDL Number: MFCD00015505 InChI Key: FSZOBSMJJFHVCQ-UHFFFAOYSA-N Synonym: 1-3,4-difluorophenyl propan-1-one,3',4'-difluoropropiophenone,3,4-difluoropropiophenone,propiophenone, 3',4'-difluoro,1-propanone, 1-3,4-difluorophenyl,acmc-1cgp0,ksc551g8r,1-3,4-difluorophenyl-1-propanone,1-propanone,1-3,4-difluorophenyl,1-3,4-difluorophenyl-1-propanone # PubChem CID: 520050 IUPAC Name: 1-(3,4-difluorophenyl)propan-1-one SMILES: CCC(=O)C1=CC=C(F)C(F)=C1