accessibility menu, dialog, popup

UserName

CH3BrNNOO

CAS RN 23056-47-5

IUPAC Name: 2-bromo-4-methyl-5-nitropyridine
Synonyms: 2-bromo-4-methyl-5-nitropyridine
Molecular Weight (g/mol): 217.02
Molecular Formula: C6H5BrN2O2
InChi Key: OSYUAHGEDKTDOX-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=NC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results