accessibility menu, dialog, popup

UserName

CH3ClNNOO

CAS RN 23056-33-9

IUPAC Name: 2-chloro-4-methyl-5-nitropyridine
Synonyms: 2-chloro-4-methyl-5-nitropyridine
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: HWZUMEVIIGNXGM-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NC=C1[N+]([O-])=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)