accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNHN

CAS RN 230301-11-8

IUPAC Name: tert-butyl 1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate
Synonyms: tert-butyl 1H,4H,6H,7H-pyrazolo[4,3-c]pyridine-5-carboxylate
Molecular Weight (g/mol): 223.28
Molecular Formula: C11H17N3O2
InChi Key: BHYPERDTLBCHAE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2=C(C1)C=NN2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)