accessibility menu, dialog, popup

UserName

OONHNH2HCl

CAS RN 22839-16-3

IUPAC Name: benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Synonyms: benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Molecular Weight (g/mol): 330.81
Molecular Formula: C18H19ClN2O2
InChi Key: DOKDMGOWZOTZRA-UHFFFAOYNA-N
SMILES: Cl.NC(CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results