CAS RN 22625-57-6
IUPAC Name:
5-(chloromethyl)-1,3-oxazolidin-2-one
Synonyms:
5-(chloromethyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol):
135.55
Molecular Formula:
C4H6ClNO2
InChi Key:
FNOZCEQRXKPZEZ-UHFFFAOYNA-N
SMILES:
ClCC1CNC(=O)O1
1
–
1
of
1
results
1
5-Chloromethyl-2-oxazolidinone 98.0+%, TCI America™
CAS: 22625-57-6 Molecular Formula: C4H6ClNO2 Molecular Weight (g/mol): 135.547 MDL Number: MFCD00005269 InChI Key: FNOZCEQRXKPZEZ-UHFFFAOYSA-N Synonym: 5-chloromethyl-2-oxazolidinone,5-chloromethyl-1,3-oxazolidin-2-one,2-oxazolidinone, 5-chloromethyl,5-chloromethyl oxazolidin-2-one,5-chloromethyloxazolidin-2-one,2-oxazolidinone,5-chloromethyl,acmc-209fxq,fnozceqrxkpzez-uhfffaoysa,5-chloromethyl-1,3-oxazolidin-2-one # PubChem CID: 90815 IUPAC Name: 5-(chloromethyl)-1,3-oxazolidin-2-one SMILES: C1C(OC(=O)N1)CCl