accessibility menu, dialog, popup

UserName

CAS RN 22591-21-5

IUPAC Name: 1,1-dichloro-3,3-dimethylbutan-2-one
Synonyms: dichloropinacolin
Molecular Weight (g/mol): 169.05
Molecular Formula: C6H10Cl2O
InChi Key: UDWZXMQIEHAAQT-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)C(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results