CAS RN 2247-91-8
IUPAC Name:
3,3,3-trifluoroprop-1-en-2-yl acetate
Synonyms:
3,3,3-trifluoroprop-1-en-2-yl acetate
Molecular Weight (g/mol):
154.09
Molecular Formula:
C5H5F3O2
InChi Key:
VOKGSDIHTCTXDS-UHFFFAOYSA-N
SMILES:
CC(=O)OC(=C)C(F)(F)F
1
–
1
of
1
results
1
1-(Trifluoromethyl)vinyl acetate, 97%
CAS: 2247-91-8 Molecular Formula: C5H5F3O2 Molecular Weight (g/mol): 154.088 MDL Number: MFCD00040844 InChI Key: VOKGSDIHTCTXDS-UHFFFAOYSA-N Synonym: 1-trifluoromethyl vinyl acetate,1-trifluoromethyl ethenyl acetate,1-propen-2-ol, 3,3,3-trifluoro-, acetate,1-propen-2-ol, 3,3,3-trifluoro-, 2-acetate,trifluoroacetone enol acetate,trifluoroacetone enolacetate,alpha-trifluoromethylvinyl acetate,acmc-1cjli,3,3,3-trifluoropropen-2-yl acetate,alpa-trifluoromethyl vinyl acetate PubChem CID: 75254 IUPAC Name: 3,3,3-trifluoroprop-1-en-2-yl acetate SMILES: CC(=O)OC(=C)C(F)(F)F