accessibility menu, dialog, popup

UserName

ZrH3CCH3OOH3CCH3OO

CAS RN 22464-99-9

IUPAC Name: λ²-zirconium(2+) bis(2-ethylhexanoate)
Synonyms: λ²-zirconium(2+) bis(2-ethylhexanoate)
Molecular Weight (g/mol): 377.64
Molecular Formula: C16H30O4Zr
InChi Key: RIYRDRZNXDRTIL-UHFFFAOYNA-L
SMILES: [Zr++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results