accessibility menu, dialog, popup

UserName

CH3NNH3CP

CAS RN 22429-12-5

IUPAC Name: 1,3-dimethyl-2-phenyl-1,3,2-diazaphospholidine
Synonyms: 1,3-dimethyl-2-phenyl-1,3,2-diazaphospholidine
Molecular Weight (g/mol): 194.22
Molecular Formula: C10H15N2P
InChi Key: CYKFZCQRONDMOZ-UHFFFAOYSA-N
SMILES: CN1CCN(C)P1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results