accessibility menu, dialog, popup

UserName

OHOO

CAS RN 22428-87-1

IUPAC Name: 1,4-dioxaspiro[4.5]decan-8-ol
Synonyms: 1,4-dioxaspiro[4.5]decan-8-ol
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: HKQTYQDNCKMNHZ-UHFFFAOYSA-N
SMILES: OC1CCC2(CC1)OCCO2
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results