CAS RN 224178-67-0
IUPAC Name:
4-(piperidin-4-yloxy)benzonitrile
Synonyms:
4-(piperidin-4-yloxy)benzonitrile
Molecular Weight (g/mol):
202.26
Molecular Formula:
C12H14N2O
InChi Key:
DRIREFRFHFBPIN-UHFFFAOYSA-N
SMILES:
N#CC1=CC=C(OC2CCNCC2)C=C1
1
–
1
of
1
results
1
Promotions Available
4-(4-Piperidinyloxy)benzonitrile, 98%
4-(4-Piperidinyloxy)benzonitrile, 98%
CAS: 224178-67-0 Molecular Formula: C12H14N2O Molecular Weight (g/mol): 202.257 MDL Number: MFCD07368391 InChI Key: DRIREFRFHFBPIN-UHFFFAOYSA-N PubChem CID: 11241109 IUPAC Name: 4-piperidin-4-yloxybenzonitrile SMILES: C1CNCCC1OC2=CC=C(C=C2)C#N