CAS RN 22325-27-5
IUPAC Name:
4,6-dimethyl-1,2-dihydropyrimidine-2-thione
Synonyms:
4,6-dimethyl-1H-pyrimidine-2-thione
Molecular Weight (g/mol):
140.2
Molecular Formula:
C6H8N2S
InChi Key:
RAFAYWADRVMWFA-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=NC(=S)N1
1
–
1
of
1
results
1
4,6-Dimethyl-2-mercaptopyrimidine 98.0+%, TCI America™
CAS: 22325-27-5 Molecular Formula: C6H8N2S Molecular Weight (g/mol): 140.204 MDL Number: MFCD00006079 InChI Key: RAFAYWADRVMWFA-UHFFFAOYSA-N Synonym: 4,6-dimethyl-2-mercaptopyrimidine,4,6-dimethyl-2 1h-pyrimidinethione,4,6-dimethylpyrimidine-2-thiol,4,6-dimethyl-2-thiopyrimidine,4,6-dimethyl-2-pyrimidinethiol,2-mercapto-4,6-dimethylpyrimidine,2 1h-pyrimidinethione, 4,6-dimethyl,4,6-dimethylpyrimidine-2-thione,4,6-dimethyl-pyrimidine-2-thiol,4,6-dimethyl-2-thiolpyrimidine PubChem CID: 673664 IUPAC Name: 4,6-dimethyl-1H-pyrimidine-2-thione SMILES: CC1=CC(=NC(=S)N1)C