CAS RN 222530-34-9
CAS RN 222530-34-9
IUPAC Name:
3-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Synonyms:
3-[(tert-butoxycarbonyl)amino]cyclohexane-1-carboxylic acid
Molecular Weight (g/mol):
243.3
Molecular Formula:
C12H21NO4
InChi Key:
JSGHMGKJNZTKGF-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC1CCCC(C1)C(O)=O
1
–
1
of
1
results
1
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid 97.0+%, TCI America™
CAS: 222530-34-9 Molecular Formula: C12H21NO4 Molecular Weight (g/mol): 243.303 MDL Number: MFCD06227743 InChI Key: JSGHMGKJNZTKGF-DTWKUNHWSA-N Synonym: (1S,3R)-3-(Boc-amino)cyclohexanecarboxylic Acid PubChem CID: 2755995 IUPAC Name: (1S,3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid SMILES: CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O