accessibility menu, dialog, popup

UserName

CAS RN 2216-32-2

H3CCH3CH3

CAS RN 2216-32-2

IUPAC Name: 4-ethylheptane
Synonyms: heptane, 4-ethyl-
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: XMROPFQWHHUFFS-UHFFFAOYSA-N
SMILES: CCCC(CC)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results