accessibility menu, dialog, popup

UserName

H3CCH3CH3CH3

CAS RN 2216-30-0

IUPAC Name: 2,5-dimethylheptane
Synonyms: heptane, 2,5-dimethyl-
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: HQZHQNKZOYIKQC-UHFFFAOYNA-N
SMILES: CCC(C)CCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results