accessibility menu, dialog, popup

UserName

CAS RN 221263-97-4

IUPAC Name: 2-[methyl(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]acetohydrazide
Synonyms: 2-[methyl(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]acetohydrazide
Molecular Weight (g/mol): 266.22
Molecular Formula: C9H10N6O4
InChi Key: IDJGYEQXPMQJKF-UHFFFAOYSA-N
SMILES: CN(CC(=O)NN)C1=CC=C(C2=NON=C12)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results