accessibility menu, dialog, popup

UserName

CAS RN 22080-96-2

IUPAC Name: 4-hydroxy-2,6-dimethoxybenzaldehyde
Synonyms: 4-hydroxy-2,6-dimethoxybenzaldehyde
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: HZWPJAZIRZFCGX-UHFFFAOYSA-N
SMILES: COC1=CC(O)=CC(OC)=C1C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results