accessibility menu, dialog, popup

UserName

CAS RN 2206-26-0

IUPAC Name: (²H₃)acetonitrile
Synonyms: (²H₃)acetonitrile
Molecular Weight (g/mol): 44.07
Molecular Formula: C2H3N
InChi Key: WEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES: [2H]C([2H])([2H])C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results