accessibility menu, dialog, popup

UserName

OO(S)BrNHOO

CAS RN 220497-47-2

IUPAC Name: (2S)-3-(2-bromophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
Synonyms: (2S)-3-(2-bromophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoate
Molecular Weight (g/mol): 465.32
Molecular Formula: C24H19BrNO4
InChi Key: GRJPAUULVKPBHU-QFIPXVFZSA-M
SMILES: [O-]C(=O)[C@H](CC1=CC=CC=C1Br)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)