accessibility menu, dialog, popup

UserName

H3CH3CCH3OOOOOCH3H3CCH3OOCH3H3CCH3OOH3CCH3CH3OOCH3H3CCH3

CAS RN 220017-47-0

IUPAC Name: {3,4,5,6-tetrakis[(2,2-dimethylpropanoyl)oxy]oxan-2-yl}methyl 2,2-dimethylpropanoate
Synonyms: {3,4,5,6-tetrakis[(2,2-dimethylpropanoyl)oxy]oxan-2-yl}methyl 2,2-dimethylpropanoate
Molecular Weight (g/mol): 600.75
Molecular Formula: C31H52O11
InChi Key: PLXCBOJERHYNCS-UHFFFAOYNA-N
SMILES: CC(C)(C)C(=O)OCC1OC(OC(=O)C(C)(C)C)C(OC(=O)C(C)(C)C)C(OC(=O)C(C)(C)C)C1OC(=O)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results