accessibility menu, dialog, popup

UserName

CAS RN 2200-70-6

IUPAC Name: octafluoro-[1,1'-biphenyl]-4,4'-diol
Synonyms: octafluoro-[1,1'-biphenyl]-4,4'-diol
Molecular Weight (g/mol): 330.13
Molecular Formula: C12H2F8O2
InChi Key: MOFZHBRFFAIMKM-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(O)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results