CAS RN 2198-54-1
IUPAC Name:
N-(3,4-dimethylphenyl)acetamide
Synonyms:
3,4-dimethylacetanilide
Molecular Weight (g/mol):
163.22
Molecular Formula:
C10H13NO
InChi Key:
UAOIEEWQVAXCFY-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=CC=C(C)C(C)=C1
1
–
1
of
1
results
1
3',4'-Dimethylacetanilide 98.0+%, TCI America™
CAS: 2198-54-1 Molecular Formula: C10H13NO Molecular Weight (g/mol): 163.22 MDL Number: MFCD00026137 InChI Key: UAOIEEWQVAXCFY-UHFFFAOYSA-N Synonym: 3',4'-dimethylacetanilide,n-3,4-dimethylphenyl acetamide,3,4-dimethylacetanilide,n-acetyl-3,4-xylidine,3',4'-acetoxylidide,acetamide, n-3,4-dimethylphenyl,3',4'-acetoxylide,acetamide, n-3,4-dimethylphenyl-9ci,4-acetamino-o-xylol,aceto-3,4-xylidide PubChem CID: 75144 IUPAC Name: N-(3,4-dimethylphenyl)acetamide SMILES: CC1=C(C=C(C=C1)NC(=O)C)C