accessibility menu, dialog, popup

UserName

CH3CH3CH3OONHOOH

CAS RN 218608-84-5

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-5-phenylpentanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-5-phenylpentanoic acid
Molecular Weight (g/mol): 293.36
Molecular Formula: C16H23NO4
InChi Key: MYWZFJXOLAXENE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results