accessibility menu, dialog, popup

UserName

CH3CH3HOCH3CH3HOCH3CH3HOCH3CH3HOHf

CAS RN 2171-99-5

IUPAC Name: tetrakis(propan-2-ol) hafnium
Synonyms: tetrakis(isopropyl alcohol) hafnium
Molecular Weight (g/mol): 418.87
Molecular Formula: C12H32HfO4
InChi Key: HMKGKDSPHSNMTM-UHFFFAOYSA-N
SMILES: [Hf].CC(C)O.CC(C)O.CC(C)O.CC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results