CAS RN 21668-77-9
IUPAC Name:
propane-1,3-disulfonate
Synonyms:
propane-1,3-disulfonate
Molecular Weight (g/mol):
202.2
Molecular Formula:
C3H6O6S2
InChi Key:
MGNVWUDMMXZUDI-UHFFFAOYSA-L
SMILES:
[O-]S(=O)(=O)CCCS([O-])(=O)=O
1
–
1
of
1
results
1
1,3-Propanedisulfonic Acid (50-60% in Water), TCI America™
CAS: 21668-77-9 Molecular Formula: C3H6O6S2 Molecular Weight (g/mol): 202.20 MDL Number: MFCD00191482 InChI Key: MGNVWUDMMXZUDI-UHFFFAOYSA-L Synonym: 1,3-propanedisulfonic acid,eprodisate,1,3-propanedisulfonic acid in water,unii-6qfp76v7s7,propane-1,3-disulfonate,eprodisate inn,1,3-propanesulfonic acid,1,3-propanedisulfonicacid,1,3-propanedisulphonic acid,1,3 propanesulfonate PubChem CID: 428573 IUPAC Name: propane-1,3-disulfonate SMILES: [O-]S(=O)(=O)CCCS([O-])(=O)=O