accessibility menu, dialog, popup

UserName

CAS RN 2155-60-4

H3COOCH2OOCH3

CAS RN 2155-60-4

IUPAC Name: 1,4-dibutyl 2-methylidenebutanedioate
Synonyms: 1,4-dibutyl 2-methylidenebutanedioate
Molecular Weight (g/mol): 242.32
Molecular Formula: C13H22O4
InChi Key: OGVXYCDTRMDYOG-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CC(=C)C(=O)OCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results